Product Details

SNP ID
rs11570576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47120357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGGATATTTGATTCCTACATAC[A/G]AGTCCCAATATGCCCATTACATTTG
Phenotype
MIM: 116946
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC27 PubMed Links

Gene Details

Gene
CDC27
Gene Name
cell division cycle 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114091.2 3134 Intron NP_001107563.1
NM_001256.4 3134 Intron NP_001247.3
NM_001293089.1 3134 Intron NP_001280018.1
NM_001293091.1 3134 Intron NP_001280020.1
XM_011525546.2 3134 UTR 3 XP_011523848.1
XM_011525547.2 3134 UTR 3 XP_011523849.1
XM_011525548.2 3134 UTR 3 XP_011523850.1
XM_011525549.2 3134 UTR 3 XP_011523851.1
XM_011525550.2 3134 UTR 3 XP_011523852.1
XM_017025481.1 3134 UTR 3 XP_016880970.1
XM_017025482.1 3134 UTR 3 XP_016880971.1
XM_017025483.1 3134 UTR 3 XP_016880972.1
XM_017025484.1 3134 UTR 3 XP_016880973.1
XM_017025485.1 3134 UTR 3 XP_016880974.1
XM_017025486.1 3134 UTR 3 XP_016880975.1
XM_017025487.1 3134 UTR 3 XP_016880976.1
XM_017025488.1 3134 UTR 3 XP_016880977.1

View Full Product Details