Product Details

SNP ID
rs12161068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26555249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTAGTAGGCTGGTGCAAAAGTAA[C/T]GCAGTTTTTGCCACTGAAAGTAATA
Phenotype
MIM: 603126
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR548J PubMed Links

Gene Details

Gene
MIR548J
Gene Name
microRNA 548j
There are no transcripts associated with this gene.

Gene
TPST2
Gene Name
tyrosylprotein sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008566.1 Intron NP_001008566.1
NM_003595.3 Intron NP_003586.3
XM_006724338.3 Intron XP_006724401.1

View Full Product Details