Product Details

SNP ID
rs11666137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33144137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGACACTGATCATTCTGGCGCTGC[C/T]CTCAGTTGGCCCCGGCCCGCCCAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR88 PubMed Links

Gene Details

Gene
WDR88
Gene Name
WD repeat domain 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173479.3 Intron NP_775750.3
XM_011526449.2 Intron XP_011524751.1
XM_011526450.2 Intron XP_011524752.1
XM_017026282.1 Intron XP_016881771.1
XM_017026283.1 Intron XP_016881772.1

View Full Product Details