Product Details

SNP ID
rs11880025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32978778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTAACTTGTACTGTGTCAACAG[A/T]GAACCTTATTAGATTCACGGTGTCA
Phenotype
MIM: 615470 MIM: 610884
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP89 PubMed Links

Gene Details

Gene
CEP89
Gene Name
centrosomal protein 89
There are no transcripts associated with this gene.

Gene
FAAP24
Gene Name
Fanconi anemia core complex associated protein 24
There are no transcripts associated with this gene.

Gene
RHPN2
Gene Name
rhophilin Rho GTPase binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033103.4 3345 UTR 3 NP_149094.3

View Full Product Details