Product Details

SNP ID
rs12727730
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.1:248036074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAAAATATTTCTCACTTACTTTG[G/T]GATTTTACATGCCATCTGAATATGG
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
OR2L13 PubMed Links
Additional Information
For this assay, SNP(s) [rs74659308] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR2L13
Gene Name
olfactory receptor family 2 subfamily L member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304535.1 Intron NP_001291464.1
NM_175911.3 Intron NP_787107.1
XM_011544169.2 Intron XP_011542471.1
Gene
OR2L2
Gene Name
olfactory receptor family 2 subfamily L member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004686.2 Intron NP_001004686.1
XM_011544158.2 Intron XP_011542460.1

View Full Product Details