Product Details

SNP ID
rs8004020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69476465 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTAATCCCAGCTACTTGGGAGGC[C/T]GAGGAGAGAGGATTGCTTGAGCTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLEKHD1 PubMed Links

Gene Details

Gene
PLEKHD1
Gene Name
pleckstrin homology and coiled-coil domain containing D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161498.1 Intron NP_001154970.1
XM_011536762.2 Intron XP_011535064.1
XM_011536763.2 Intron XP_011535065.1
XM_017021290.1 Intron XP_016876779.1

View Full Product Details