Product Details

SNP ID
rs11860456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55846568 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGGGGGATGGTGCTGGTCCAAAA[A/C]TCCACCCGCGGTTCTTTGAGTCTCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CES5A PubMed Links

Gene Details

Gene
CES5A
Gene Name
carboxylesterase 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143685.1 1760 Missense Mutation GAG,GAT E537D NP_001137157.1
NM_001190158.1 1760 Missense Mutation GAG,GAT E566D NP_001177087.1
NM_145024.2 1760 Missense Mutation GAG,GAT E487D NP_659461.1

View Full Product Details