Product Details

SNP ID
rs11860488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55846622 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGAGGCTCATGTTCAAGTCCAGCTG[C/G]AGGTACTGCTCAGTCAGATTATAAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CES5A PubMed Links

Gene Details

Gene
CES5A
Gene Name
carboxylesterase 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143685.1 1706 Silent Mutation CTC,CTG L519L NP_001137157.1
NM_001190158.1 1706 Silent Mutation CTC,CTG L548L NP_001177087.1
NM_145024.2 1706 Silent Mutation CTC,CTG L469L NP_659461.1

View Full Product Details