Product Details
- SNP ID
-
rs10844566
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:33376552 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTGTTTAAAAAAACATTTCATATG[C/G]AAAGAATTACAACATAGTAAAACAC
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SYT10
PubMed Links
Gene Details
- Gene
- SYT10
- Gene Name
- synaptotagmin 10
View Full Product Details