Product Details
- SNP ID
-
rs12228193
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:32796000 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGCCTGGCTTTACATCCTGTTTGC[C/T]GCCATGTTGCACATGATGGTCATGA
- Phenotype
-
MIM: 602861
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PKP2
PubMed Links
Gene Details
- Gene
- PKP2
- Gene Name
- plakophilin 2
View Full Product Details