Product Details

SNP ID
rs12452844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76463161 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGTGCCCGGGATGGGGTGGGGGCC[A/G]TGGGGAGGTTTTCTCCAACCCCCGG
Phenotype
MIM: 600950 MIM: 614404
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AANAT PubMed Links

Gene Details

Gene
AANAT
Gene Name
aralkylamine N-acetyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001088.2 Intron NP_001079.1
NM_001166579.1 Intron NP_001160051.1
XM_017024259.1 Intron XP_016879748.1
Gene
RHBDF2
Gene Name
rhomboid 5 homolog 2
There are no transcripts associated with this gene.

Gene
UBE2O
Gene Name
ubiquitin conjugating enzyme E2 O
There are no transcripts associated with this gene.

View Full Product Details