Product Details

SNP ID
rs12756225
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:180913259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCGAGGGATGGAGGGGACAGAG[G/T]CGGCGGCGGCCAAACCCGCGGGCGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1614 PubMed Links

Gene Details

Gene
KIAA1614
Gene Name
KIAA1614
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020950.1 83 Missense Mutation GCG,TCG A6S NP_066001.1

View Full Product Details