Product Details

SNP ID
rs12754176
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:198160123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTTACAGTGTTTCCAGAACTAA[A/C]CACTATTTTCCTAGGAACTATCTGA
Phenotype
MIM: 606848
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NEK7 PubMed Links

Gene Details

Gene
NEK7
Gene Name
NIMA related kinase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133494.2 Intron NP_598001.1
XM_011509209.1 Intron XP_011507511.1
XM_017000344.1 Intron XP_016855833.1
XM_017000345.1 Intron XP_016855834.1
XM_017000346.1 Intron XP_016855835.1
XM_017000347.1 Intron XP_016855836.1
XM_017000348.1 Intron XP_016855837.1

View Full Product Details