Product Details

SNP ID
rs12816175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:22194779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATGCATAGCCCACTTTCCAAGG[G/T]TGAGGGTCAAAGTGACACTTGCTGC
Phenotype
MIM: 601123
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ST8SIA1 PubMed Links

Gene Details

Gene
ST8SIA1
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304450.1 8326 UTR 3 NP_001291379.1
NM_003034.3 8326 UTR 3 NP_003025.1

View Full Product Details