Product Details

SNP ID
rs11615386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120716736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCCGCCTCCGGACAGTCGGGG[A/C]TACGTGAGTACCATTACTACCTGAG
Phenotype
MIM: 606885
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACADS PubMed Links

Gene Details

Gene
ACADS
Gene Name
acyl-CoA dehydrogenase, C-2 to C-3 short chain
There are no transcripts associated with this gene.

Gene
MIR4700
Gene Name
microRNA 4700
There are no transcripts associated with this gene.

Gene
UNC119B
Gene Name
unc-119 lipid binding chaperone B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080533.2 507 Missense Mutation GAT,GCT D156A NP_001074002.1

View Full Product Details