Product Details

SNP ID
rs12207589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87168483 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTTATATATTAGCTTCGTAATAG[G/T]TTTTTTTGAGGTCGAGTTTCGCTCT
Phenotype
MIM: 616213
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF292 PubMed Links

Gene Details

Gene
ZNF292
Gene Name
zinc finger protein 292
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015021.1 Intron NP_055836.1
XM_005248697.4 Intron XP_005248754.1
XM_011535625.2 Intron XP_011533927.1
XM_017010575.1 Intron XP_016866064.1
XM_017010576.1 Intron XP_016866065.1
XM_017010577.1 Intron XP_016866066.1
XM_017010578.1 Intron XP_016866067.1
XM_017010579.1 Intron XP_016866068.1
XM_017010580.1 Intron XP_016866069.1
XM_017010581.1 Intron XP_016866070.1
XM_017010582.1 Intron XP_016866071.1
XM_017010583.1 Intron XP_016866072.1
XM_017010584.1 Intron XP_016866073.1

View Full Product Details