Product Details

SNP ID
rs11685727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:151417244 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGCTTGGGTTGTTTAGGAAGGCA[A/G]TCAGTGTTTAGCAGGGAATAGAGTA
Phenotype
MIM: 608952
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RIF1 PubMed Links

Gene Details

Gene
RIF1
Gene Name
replication timing regulatory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177663.1 Intron NP_001171134.1
NM_001177664.1 Intron NP_001171135.1
NM_001177665.1 Intron NP_001171136.1
NM_018151.4 Intron NP_060621.3
XM_005246665.3 Intron XP_005246722.1
XM_011511395.2 Intron XP_011509697.1
XM_017004422.1 Intron XP_016859911.1
XM_017004423.1 Intron XP_016859912.1
XM_017004424.1 Intron XP_016859913.1
XM_017004425.1 Intron XP_016859914.1

View Full Product Details