Product Details

SNP ID
rs12566579
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:16472300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGTCTTGCTTTCCTTTGGCTAA[A/G]GTTTGCATGATATATCTTTTCCCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CROCCP3 PubMed Links

Gene Details

Gene
CROCCP3
Gene Name
ciliary rootlet coiled-coil, rootletin pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details