Product Details

SNP ID
rs13186308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34933935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCATCACGCTGTCCTTCCCATC[C/T]TAGTTTATGATGTTGGGAGCAGTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRIX1 PubMed Links

Gene Details

Gene
BRIX1
Gene Name
BRX1, biogenesis of ribosomes
There are no transcripts associated with this gene.

Gene
DNAJC21
Gene Name
DnaJ heat shock protein family (Hsp40) member C21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012339.2 Intron NP_001012339.2
NM_194283.3 Intron NP_919259.3
XM_005248249.4 Intron XP_005248306.2
XM_005248250.3 Intron XP_005248307.2
XM_011513965.2 Intron XP_011512267.2
XM_011513966.2 Intron XP_011512268.2

View Full Product Details