Product Details
- SNP ID
-
rs11260566
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
41
- Location
-
Chr.1:1242865 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTGCAGCAGCCCCTGCACCTGTAG[C/T]GTGAAGACCCTGCTGTTGCTCTCCA
- Phenotype
-
MIM: 615291
MIM: 616593
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
B3GALT6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74511182] are located under a probe and SNP(s) [rs77651834] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- B3GALT6
- Gene Name
- beta-1,3-galactosyltransferase 6
There are no transcripts associated with this gene.
- Gene
- FAM132A
- Gene Name
- family with sequence similarity 132 member A
View Full Product Details