Product Details

SNP ID
hCV31638092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:111465320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTACTGTTAATTAAGATACAAAA[C/T]AGAGGAGAAGAAGCAGGTTTTAAGG
Phenotype
MIM: 603270
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP5F1 PubMed Links

Gene Details

Gene
ATP5F1
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1
There are no transcripts associated with this gene.

Gene
C1orf162
Gene Name
chromosome 1 open reading frame 162
There are no transcripts associated with this gene.

View Full Product Details