Product Details

SNP ID
rs12085442
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:111333864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCATCTGCAATCAGCCCTCTTGC[G/T]GATGTTCAGAACCTCATAACCAGAA
Phenotype
MIM: 614234
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PIFO PubMed Links

Gene Details

Gene
PIFO
Gene Name
primary cilia formation
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300831.1 Intron NP_001287760.1
NM_181643.5 Intron NP_857594.2
XM_005270472.1 Intron XP_005270529.1
XM_017000322.1 Intron XP_016855811.1

View Full Product Details