Product Details

SNP ID
rs13344123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41311289 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTAGGACAAGCTACTCAGGAGACT[A/C]AGGTGGGAGTGTCACTCGAATCCTT
Phenotype
MIM: 605800
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC97 PubMed Links

Gene Details

Gene
CCDC97
Gene Name
coiled-coil domain containing 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052848.1 Intron NP_443080.1
XM_005259377.4 Intron XP_005259434.1
XM_017027442.1 Intron XP_016882931.1
Gene
HNRNPUL1
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
There are no transcripts associated with this gene.

View Full Product Details