Product Details

SNP ID
rs12447825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71904653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACTCCTGAGCTCCAGCGATCCT[A/C]CCACATCAGCCTCCCAAAGTGCTGG
Phenotype
MIM: 616434
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IST1 PubMed Links

Gene Details

Gene
IST1
Gene Name
IST1, ESCRT-III associated factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270975.1 Intron NP_001257904.1
NM_001270976.1 Intron NP_001257905.1
NM_001270977.1 Intron NP_001257906.1
NM_001270978.1 Intron NP_001257907.1
NM_001270979.1 Intron NP_001257908.1
NM_014761.3 Intron NP_055576.2

View Full Product Details