Product Details
- SNP ID
-
rs12275284
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:118259012 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAATGGCCTAATACACGGTTATGAT[A/T]AAAAAAAAAAAAGACAATGAGTGTT
- Phenotype
-
MIM: 604873
MIM: 611707
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MPZL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139129059] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MPZL2
- Gene Name
- myelin protein zero like 2
- Gene
- MPZL3
- Gene Name
- myelin protein zero like 3
There are no transcripts associated with this gene.
View Full Product Details