Product Details

SNP ID
rs11216906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118573229 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGACCACTGGGAAAATGTGAGCA[A/G]GTTCCAGAGAACACAGGGTTTGCCT
Phenotype
MIM: 600820
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARCN1 PubMed Links

Gene Details

Gene
ARCN1
Gene Name
archain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142281.1 2396 Intron NP_001135753.1
NM_001655.4 2396 Intron NP_001646.2
XM_005271542.3 2396 Intron XP_005271599.1
Gene
IFT46
Gene Name
intraflagellar transport 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168618.1 2396 Intron NP_001162089.1
NM_020153.3 2396 Intron NP_064538.3
XM_011542905.2 2396 UTR 5 XP_011541207.1
XM_011542906.2 2396 UTR 5 XP_011541208.1
XM_017018017.1 2396 Intron XP_016873506.1
XM_017018018.1 2396 Intron XP_016873507.1

View Full Product Details