Product Details

SNP ID
rs11216909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118579031 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTGCTACAAGTTGTATAGCACTT[C/T]TGAAGTACAGTTTCATTATAGTCAT
Phenotype
MIM: 600820
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARCN1 PubMed Links

Gene Details

Gene
ARCN1
Gene Name
archain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142281.1 Intron NP_001135753.1
NM_001655.4 Intron NP_001646.2
XM_005271542.3 Intron XP_005271599.1
Gene
IFT46
Gene Name
intraflagellar transport 46
There are no transcripts associated with this gene.

View Full Product Details