Product Details
- SNP ID
-
rs11040886
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:6398532 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTCCAAGCGGAGCAGTCAAGGGTC[A/G]CAGGCAGTTGATCTGGCATAGGGAG
- Phenotype
-
MIM: 602709
MIM: 607608
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APBB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114912691] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APBB1
- Gene Name
- amyloid beta precursor protein binding family B member 1
- Gene
- SMPD1
- Gene Name
- sphingomyelin phosphodiesterase 1
There are no transcripts associated with this gene.
View Full Product Details