Product Details

SNP ID
rs11665815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1010904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAGATCATGGAATGCTGCGGGA[A/G]GGAGAGTGGGAGTCAGGACGGGCCC
Phenotype
MIM: 606651 MIM: 611011
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIN3B PubMed Links

Gene Details

Gene
GRIN3B
Gene Name
glutamate ionotropic receptor NMDA type subunit 3B
There are no transcripts associated with this gene.

Gene
LOC105372235
Gene Name
uncharacterized LOC105372235
There are no transcripts associated with this gene.

Gene
TMEM259
Gene Name
transmembrane protein 259
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033026.1 Intron NP_001028198.1
NM_033420.3 Intron NP_219488.1
XM_005259675.2 Intron XP_005259732.1
XM_005259676.2 Intron XP_005259733.1
XM_005259677.2 Intron XP_005259734.1
XM_005259678.2 Intron XP_005259735.1
XM_017027457.1 Intron XP_016882946.1

View Full Product Details