Product Details

SNP ID
rs12745610
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:224385899 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACTTCACATGTGCACGAGACTTT[C/T]AGTAAAAATGACAAGATCCTAGAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNIH4 PubMed Links

Gene Details

Gene
CNIH4
Gene Name
cornichon family AMPA receptor auxiliary protein 4
There are no transcripts associated with this gene.

Gene
WDR26
Gene Name
WD repeat domain 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001115113.2 5179 UTR 3 NP_001108585.2
NM_025160.6 5179 UTR 3 NP_079436.4
XM_005273273.4 5179 UTR 3 XP_005273330.1
XM_006711817.3 5179 UTR 3 XP_006711880.1
XM_011544281.2 5179 UTR 3 XP_011542583.1
XM_011544282.2 5179 UTR 3 XP_011542584.1
XM_017002396.1 5179 UTR 3 XP_016857885.1

View Full Product Details