Product Details
- SNP ID
-
rs12092566
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:224392797 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCTTCTACAGTCTGTACCACAAA[A/G]ATAAAAGTGTAGGGAATATACTTTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4742
PubMed Links
Gene Details
- Gene
- MIR4742
- Gene Name
- microRNA 4742
There are no transcripts associated with this gene.
- Gene
- WDR26
- Gene Name
- WD repeat domain 26
View Full Product Details