Product Details

SNP ID
rs12075333
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:240007677 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCATTCTGCTGAGGAACTGAAAG[C/T]GGGTTTCCCACTCCAATGTCTTAGT
Phenotype
MIM: 606373
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FMN2 PubMed Links

Gene Details

Gene
FMN2
Gene Name
formin 2
There are no transcripts associated with this gene.

Gene
RPS7P5
Gene Name
ribosomal protein S7 pseudogene 5
There are no transcripts associated with this gene.

View Full Product Details