Product Details

SNP ID
rs12981377
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54862831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTTTTTTTTTTTTTGTATAG[G/T]GCTTCTGATGAGCTTTTTTTTAAAA
Phenotype
MIM: 604947
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
KIR3DL2 PubMed Links

Gene Details

Gene
KIR3DL2
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242867.1 Intron NP_001229796.1
NM_006737.3 Intron NP_006728.2
XM_011526940.2 Intron XP_011525242.1
XM_017026784.1 Intron XP_016882273.1

View Full Product Details