Product Details

SNP ID
rs12144983
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:244980425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGACAATCATATTTGTTGTCTA[A/G]AAAAATCACTTGGGCAAGTGTGCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFCAB2 PubMed Links

Gene Details

Gene
EFCAB2
Gene Name
EF-hand calcium binding domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143943.1 Intron NP_001137415.1
NM_001290327.1 Intron NP_001277256.1
NM_032328.3 Intron NP_115704.1
XM_011544300.2 Intron XP_011542602.1
XM_011544301.1 Intron XP_011542603.1
XM_011544302.2 Intron XP_011542604.1
XM_017002538.1 Intron XP_016858027.1
XM_017002539.1 Intron XP_016858028.1
XM_017002540.1 Intron XP_016858029.1
XM_017002541.1 Intron XP_016858030.1
XM_017002542.1 Intron XP_016858031.1
XM_017002543.1 Intron XP_016858032.1
XM_017002544.1 Intron XP_016858033.1
XM_017002545.1 Intron XP_016858034.1
Gene
LOC101928068
Gene Name
uncharacterized LOC101928068
There are no transcripts associated with this gene.

View Full Product Details