Product Details

SNP ID
rs13337834
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2347065 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCATTGTGGTTTAAATTTGTATT[A/T]CCCTAATGATTAATGATGTTGAGCA
Phenotype
MIM: 601615
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
ABCA17P PubMed Links

Gene Details

Gene
ABCA17P
Gene Name
ATP binding cassette subfamily A member 17, pseudogene
There are no transcripts associated with this gene.

Gene
ABCA3
Gene Name
ATP binding cassette subfamily A member 3
There are no transcripts associated with this gene.

View Full Product Details