Product Details

SNP ID
rs7092587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:6149278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGAGGGACAAGGGAATAGCAGG[C/T]GAAAGTTCTAATAATAAATAACAAT
Phenotype
MIM: 605319
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR3155A PubMed Links

Gene Details

Gene
MIR3155A
Gene Name
microRNA 3155a
There are no transcripts associated with this gene.

Gene
MIR3155B
Gene Name
microRNA 3155b
There are no transcripts associated with this gene.

Gene
PFKFB3
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145443.2 Intron NP_001138915.1
NM_001282630.2 Intron NP_001269559.1
NM_001314063.1 Intron NP_001300992.1
NM_001323016.1 Intron NP_001309945.1
NM_001323017.1 Intron NP_001309946.1
NM_004566.3 Intron NP_004557.1
XM_005252464.1 Intron XP_005252521.1
XM_011519493.1 Intron XP_011517795.1
XM_017016325.1 Intron XP_016871814.1
XM_017016326.1 Intron XP_016871815.1
XM_017016327.1 Intron XP_016871816.1
XM_017016328.1 Intron XP_016871817.1
XM_017016329.1 Intron XP_016871818.1

View Full Product Details