Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015155.2 | 4249 | UTR 3 | NP_055970.1 | ||
XM_005252432.3 | 4249 | Intron | XP_005252489.1 | ||
XM_005252435.3 | 4249 | Intron | XP_005252492.1 | ||
XM_011519435.2 | 4249 | Intron | XP_011517737.1 | ||
XM_017015987.1 | 4249 | UTR 3 | XP_016871476.1 | ||
XM_017015988.1 | 4249 | UTR 3 | XP_016871477.1 | ||
XM_017015989.1 | 4249 | UTR 3 | XP_016871478.1 | ||
XM_017015990.1 | 4249 | Intron | XP_016871479.1 | ||
XM_017015991.1 | 4249 | Intron | XP_016871480.1 | ||
XM_017015992.1 | 4249 | UTR 3 | XP_016871481.1 | ||
XM_017015993.1 | 4249 | UTR 3 | XP_016871482.1 | ||
XM_017015994.1 | 4249 | UTR 3 | XP_016871483.1 | ||
XM_017015995.1 | 4249 | Intron | XP_016871484.1 |