Product Details

SNP ID
rs10982986
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:115891387 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAAAATATTTCAAATATATGTTG[A/G]TAAGTGCTCTATTCAACCTTGTGCA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LINC00474 PubMed Links

Gene Details

Gene
LINC00474
Gene Name
long intergenic non-protein coding RNA 474
There are no transcripts associated with this gene.

View Full Product Details