Product Details

SNP ID
rs9657933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120325268 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAGACAGAGTTGCTCTGTTACCC[A/C]GGCTGGACAGCAGTGACACGACCTT
Phenotype
MIM: 172410
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PLA2G1B PubMed Links

Gene Details

Gene
PLA2G1B
Gene Name
phospholipase A2 group IB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000928.2 Intron NP_000919.1

View Full Product Details