Product Details

SNP ID
rs12810628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120325883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTGTCCAGTTCATCCACGGGGG[C/T]GCCTGAGCCCCCCAAGCCACAGTAG
Phenotype
MIM: 172410
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G1B PubMed Links

Gene Details

Gene
PLA2G1B
Gene Name
phospholipase A2 group IB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000928.2 208 Missense Mutation CCC,TCC P58S NP_000919.1

View Full Product Details