Product Details

SNP ID
rs3743081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78170787 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGCACAGTCCTGTGAGTTAAATG[C/T]CCACTACTCTCTGCACTGGTCCCAC
Phenotype
MIM: 614362 MIM: 601149
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSBG1 PubMed Links

Gene Details

Gene
ACSBG1
Gene Name
acyl-CoA synthetase bubblegum family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199377.1 2852 Intron NP_001186306.1
NM_015162.4 2852 Intron NP_055977.3
XM_011521390.2 2852 UTR 3 XP_011519692.1
XM_011521391.2 2852 Intron XP_011519693.2
XM_017022025.1 2852 Intron XP_016877514.1
XM_017022026.1 2852 UTR 3 XP_016877515.1
Gene
IDH3A
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005530.2 2852 Intron NP_005521.1
XM_005254336.2 2852 Intron XP_005254393.1
XM_017022118.1 2852 Intron XP_016877607.1
XM_017022119.1 2852 Intron XP_016877608.1
XM_017022120.1 2852 Intron XP_016877609.1

View Full Product Details