Product Details

SNP ID
rs12471620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24790654 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTCAAAAGGCCAAAAAAGGTTTC[C/G]GGAGTCCTCTTGCCTGAAAGTGGAG
Phenotype
MIM: 611504
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CENPO PubMed Links

Gene Details

Gene
CENPO
Gene Name
centromere protein O
There are no transcripts associated with this gene.

Gene
PTRHD1
Gene Name
peptidyl-tRNA hydrolase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013663.1 Intron NP_001013685.1

View Full Product Details