Product Details

SNP ID
rs13440475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71580203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCCAGTGGGATGTGGGGGCGGT[C/T]GAGCCCTGATACGTGGAAGACTGGC
Phenotype
MIM: 300369 MIM: 300255
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACRC PubMed Links

Gene Details

Gene
ACRC
Gene Name
acidic repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052957.4 Intron NP_443189.1
XM_006724716.3 Intron XP_006724779.1
XM_011531063.2 Intron XP_011529365.1
XM_011531064.1 Intron XP_011529366.1
XM_011531066.2 Intron XP_011529368.1
XM_011531067.2 Intron XP_011529369.1
XM_017029965.1 Intron XP_016885454.1
Gene
OGT
Gene Name
O-linked N-acetylglucosamine (GlcNAc) transferase
There are no transcripts associated with this gene.

View Full Product Details