Product Details

SNP ID
rs12087292
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:35277190 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACATCTCCGATAAATTAATCAGAG[A/C]CCATGAGGTAAAATGTCATCATCAA
Phenotype
MIM: 613568
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZMYM4 PubMed Links

Gene Details

Gene
ZMYM4
Gene Name
zinc finger MYM-type containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005095.2 Intron NP_005086.2
XM_005271328.4 Intron XP_005271385.1
XM_005271331.2 Intron XP_005271388.1
XM_011542421.2 Intron XP_011540723.1
XM_011542423.2 Intron XP_011540725.1
XM_011542425.2 Intron XP_011540727.1
XM_017002803.1 Intron XP_016858292.1
XM_017002804.1 Intron XP_016858293.1
XM_017002805.1 Intron XP_016858294.1

View Full Product Details