Product Details

SNP ID
rs10794535
Assay Type
Functionally tested
NCBI dbSNP Submissions
55
Location
Chr.1:26531463 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAGGGCTGGGAGAGGCTGGGGAG[A/T]GACTTTATGCTGATAGGGAGGGTGG
Phenotype
MIM: 601684
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RPS6KA1 PubMed Links

Gene Details

Gene
RPS6KA1
Gene Name
ribosomal protein S6 kinase A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006665.1 Intron NP_001006666.1
NM_002953.3 Intron NP_002944.2
XM_005245966.4 Intron XP_005246023.1

View Full Product Details