Product Details

SNP ID
rs12481037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25613052 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATGAAAAATATCATCACACGTAA[C/T]AAGAATGTTTTATGAAAATTAAACA
Phenotype
MIM: 610763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NANP PubMed Links

Gene Details

Gene
NANP
Gene Name
N-acetylneuraminic acid phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152667.2 3684 UTR 3 NP_689880.1

View Full Product Details