Product Details
- SNP ID
-
rs13428985
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:171696095 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGGCATTCTTTGTGTATTTTGTAT[A/G]TTTTAGGGAGGACTTTTGTCTGTTT
- Phenotype
-
MIM: 603331
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DYNC1I2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78293586] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DYNC1I2
- Gene Name
- dynein cytoplasmic 1 intermediate chain 2
View Full Product Details