Product Details

SNP ID
rs13184447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178943642 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCTTTTTCTTTTATGACATTTGG[C/T]TTTTTTACCAATGAAATTATTCAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF454 PubMed Links

Gene Details

Gene
ZNF454
Gene Name
zinc finger protein 454
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178089.2 Intron NP_001171560.1
NM_001178090.2 Intron NP_001171561.1
NM_001323306.1 Intron NP_001310235.1
NM_001323307.1 Intron NP_001310236.1
NM_001323308.1 Intron NP_001310237.1
NM_001323309.1 Intron NP_001310238.1
NM_001323310.1 Intron NP_001310239.1
NM_182594.3 Intron NP_872400.2

View Full Product Details