Product Details

SNP ID
rs13298231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37591394 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTTTTGGATTTCAAAGGTCAGCA[C/T]TTTTGAAAGGGAAGGAACCCGGCCG
Phenotype
MIM: 616169
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOMM5 PubMed Links

Gene Details

Gene
TOMM5
Gene Name
translocase of outer mitochondrial membrane 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001790.2 Intron NP_001001790.1
NM_001134484.1 Intron NP_001127956.1
NM_001134485.1 Intron NP_001127957.1

View Full Product Details