Product Details

SNP ID
rs10980999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:111662075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCCACCGCAGGGCGGCGAATT[A/G]GTGATGGATTGGTAGGATGAGAGCG
Phenotype
MIM: 604389
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC25 PubMed Links

Gene Details

Gene
DNAJC25
Gene Name
DnaJ heat shock protein family (Hsp40) member C25
There are no transcripts associated with this gene.

Gene
DNAJC25-GNG10
Gene Name
DNAJC25-GNG10 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004125.3 Intron NP_004116.2
Gene
GNG10
Gene Name
G protein subunit gamma 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017998.3 Intron NP_001017998.1
NM_001198664.1 Intron NP_001185593.1

View Full Product Details